ENA333559108 Enamine

3-(4-bromo-3,5-dimethyl-1H-pyrazol-1-yl)-2-methylpropanoic acid

  • MDL number MFCD04967205
  • Cas Number : 890603-20-0
  • Molecular Weight : 261.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(4-bromo-3,5-dimethyl-1H-pyrazol-1-yl)-2-methylpropanoic acid
Inchi Code 1S/C9H13BrN2O2/c1-5(9(13)14)4-12-7(3)8(10)6(2)11-12/h5H,4H2,1-3H3,(H,13,14)
Inchi Key BZLIXBYMKOSUCJ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-83658

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83658
Availability details for
Quantity
  • Availability
  • Add to Cart