ENA333559112 Enamine

2-[(3-methyl-4-nitro-1H-pyrazol-5-yl)sulfanyl]acetic acid

  • MDL number MFCD00464488
  • Cas Number : 879565-31-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83662

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83662
Availability details for
Quantity
  • Availability
  • Add to Cart