ENA333559115 Enamine

1-[(1H-pyrazol-1-yl)methyl]-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD04967756
  • Cas Number : 1005631-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-83665

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83665
Availability details for
Quantity
  • Availability
  • Add to Cart