ENA333559118 Enamine

1-[(4-bromo-1H-pyrazol-1-yl)methyl]-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD04049359
  • Cas Number : 1001755-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83668

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83668
Availability details for
Quantity
  • Availability
  • Add to Cart