ENA333559137 Enamine

1-(4-fluorophenoxymethyl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD04967923
  • Cas Number : 1004193-27-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83687
Availability details for
Quantity
  • Availability
  • Add to Cart