ENA333559142 Enamine

1-[(4-bromo-3,5-dimethyl-1H-pyrazol-1-yl)methyl]-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD04969101
  • Cas Number : 1004194-36-8
  • Molecular Weight : 299.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-83692

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83692
Availability details for
Quantity
  • Availability
  • Add to Cart