ENA333559146 Enamine

2-[(4-nitro-1H-pyrazol-1-yl)methyl]benzoic acid

  • MDL number MFCD04969107
  • Cas Number : 925145-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83696

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83696
Availability details for
Quantity
  • Availability
  • Add to Cart