ENA333559150 Enamine

3-(1H-indol-3-yl)butanoic acid

  • MDL number MFCD00081139
  • Cas Number : 3569-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83700

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83700
Availability details for
Quantity
  • Availability
  • Add to Cart