ENA333559151 Enamine

3-(3-methyl-2,5-dioxopyrrolidin-1-yl)benzoic acid

  • MDL number MFCD04969729
  • Cas Number : 832739-55-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83701

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83701
Availability details for
Quantity
  • Availability
  • Add to Cart