ENA333559154 Enamine

5-(1,3-dimethyl-1H-pyrazol-4-yl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD04969774
  • Cas Number : 1030620-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83705
Availability details for
Quantity
  • Availability
  • Add to Cart