ENA333559175 Enamine

3-(naphthalene-2-sulfonyl)propanoic acid

  • MDL number MFCD00222810
  • Cas Number : 32385-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-83726

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83726
Availability details for
Quantity
  • Availability
  • Add to Cart