ENA333559178 Enamine

1-ethyl-5-(trifluoromethyl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD04970207
  • Cas Number : 1004643-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83729

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83729
Availability details for
Quantity
  • Availability
  • Add to Cart