ENA333559180 Enamine

5-(1-ethyl-5-methyl-1H-pyrazol-4-yl)-1,2-oxazole-3-carboxylic acid

  • MDL number MFCD04970282
  • Cas Number : 957487-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83731

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83731
Availability details for
Quantity
  • Availability
  • Add to Cart