ENA333559181 Enamine

4-methyl-5-(5-methylfuran-2-yl)thiophene-2-carboxylic acid

  • MDL number MFCD04970817
  • Cas Number : 832740-66-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83732

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83732
Availability details for
Quantity
  • Availability
  • Add to Cart