ENA333559250 Enamine

5-(4-methoxyphenyl)-N-methyl-1H-pyrazol-3-amine

  • MDL number MFCD19982530
  • Cas Number : 101476-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83772

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83772
Availability details for
Quantity
  • Availability
  • Add to Cart