ENA333559263 Enamine

3-methanesulfonylcyclohexan-1-amine hydrochloride

  • MDL number MFCD19982565
  • Cas Number : 1334147-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83964

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83964
Availability details for
Quantity
  • Availability
  • Add to Cart