ENA333559268 Enamine

2-(benzyloxy)-1-phenylethan-1-amine hydrochloride

  • MDL number MFCD19982577
  • Cas Number : 741260-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84083

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-84083
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart