ENA333559271 Enamine

N,5-dimethyl-1H-pyrazol-3-amine

  • MDL number MFCD17282966
  • Cas Number : 73844-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84180

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-84180
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart