ENA333559272 Enamine

2-[2-(difluoromethoxy)phenyl]ethan-1-amine hydrochloride

  • MDL number MFCD19982585
  • Cas Number : 1334148-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84201

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84201
Availability details for
Quantity
  • Availability
  • Add to Cart