ENA333559279 Enamine

7-bromo-3H-imidazo[4,5-c]pyridine-2-thiol

  • MDL number MFCD19982592
  • Cas Number : 120759-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84251

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84251
Availability details for
Quantity
  • Availability
  • Add to Cart