ENA333559286 Enamine

2-(1,3-thiazol-2-yl)butan-2-amine hydrochloride

  • MDL number MFCD19982601
  • Cas Number : 1334147-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84370

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84370
Availability details for
Quantity
  • Availability
  • Add to Cart