ENA333559318 Enamine

3-{ethyl[(8-methyl-4-oxo-3,4-dihydroquinazolin-2-yl)methyl]amino}propanoic acid

  • MDL number MFCD18473323
  • Cas Number : 1179747-61-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83859

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83859
Availability details for
Quantity
  • Availability
  • Add to Cart