ENA333559325 Enamine

5-(1,3-dioxaindan-5-yl)-1,2-oxazole-3-carboxylic acid

  • MDL number MFCD04113953
  • Cas Number : 668971-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83744

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83744
Availability details for
Quantity
  • Availability
  • Add to Cart