ENA333559326 Enamine

2-amino-1-(2,6-difluorophenyl)ethan-1-ol hydrochloride

  • MDL number MFCD09831936
  • Cas Number : 1333642-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204...206
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83876

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83876
Availability details for
Quantity
  • Availability
  • Add to Cart