ENA333559332 Enamine

methyl 2-amino-4-fluoro-5-(4-fluorophenoxy)benzoate

  • MDL number MFCD19982559
  • Cas Number : 1333657-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-83883

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83883
Availability details for
Quantity
  • Availability
  • Add to Cart