ENA333559335 Enamine

methyl 3-(4-methoxyphenyl)-2-methyl-3-oxopropanoate

  • MDL number MFCD16076972
  • Cas Number : 86109-34-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83886

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83886
Availability details for
Quantity
  • Availability
  • Add to Cart