ENA333559336 Enamine

1-(3-methoxyphenyl)propan-1-ol

  • MDL number MFCD11521209
  • Cas Number : 52956-27-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83887

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83887
Availability details for
Quantity
  • Availability
  • Add to Cart