ENA333559338 Enamine

3,5-dimethyl-1H-pyrazole-1-carbothioamide

  • MDL number MFCD00180878
  • Cas Number : 1124-15-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83891

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83891
Availability details for
Quantity
  • Availability
  • Add to Cart