ENA333559343 Enamine

4-methyl-2-phenylpentanoic acid

  • MDL number MFCD00230422
  • Cas Number : 14320-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77-79
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-83901

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83901
Availability details for
Quantity
  • Availability
  • Add to Cart