ENA333559345 Enamine

4-methoxy-3-[(piperidin-1-yl)methyl]benzoic acid

  • MDL number MFCD05667174
  • Cas Number : 832739-97-6
  • Molecular Weight : 249.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-83746

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83746
Availability details for
Quantity
  • Availability
  • Add to Cart