ENA333559350 Enamine

8-chloro-3-iodoquinoline

  • MDL number MFCD11044980
  • Cas Number : 847727-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97...99
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83917

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83917
Availability details for
Quantity
  • Availability
  • Add to Cart