ENA333559351 Enamine

(2S)-2-(2-methoxyacetamido)-3-methylbutanoic acid

  • MDL number MFCD14588533
  • Cas Number : 257638-51-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83918

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83918
Availability details for
Quantity
  • Availability
  • Add to Cart