ENA333559359 Enamine

2-(4-tert-butylphenyl)cyclopropane-1-carboxylic acid

  • MDL number MFCD02056423
  • Cas Number : 445029-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83734

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83734
Availability details for
Quantity
  • Availability
  • Add to Cart