ENA333559394 Enamine

4-ethyl-5-(4-methylphenyl)thiophene-2-carboxylic acid

  • MDL number MFCD05861654
  • Cas Number : 869950-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83749

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83749
Availability details for
Quantity
  • Availability
  • Add to Cart