ENA333559397 Enamine

1-[(4-fluorophenyl)methyl]piperidine-3-carboxylic acid hydrochloride

  • MDL number MFCD06800730
  • Cas Number : 1185303-78-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-83751

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83751
Availability details for
Quantity
  • Availability
  • Add to Cart