ENA333559399 Enamine

1-amino-2,5-dimethylhexan-2-ol

  • MDL number MFCD12169425
  • Cas Number : 1155915-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84051

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84051
Availability details for
Quantity
  • Availability
  • Add to Cart