ENA333559402 Enamine

3-methyl-3-(2-methylpropyl)pyrrolidine-2,5-dione

  • MDL number MFCD11207843
  • Cas Number : 1094231-95-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84054

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84054
Availability details for
Quantity
  • Availability
  • Add to Cart