ENA333559411 Enamine

1-(4-chlorophenyl)-3-methylbutan-1-one

  • MDL number MFCD00032766
  • Cas Number : 71573-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84066

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84066
Availability details for
Quantity
  • Availability
  • Add to Cart