ENA333559415 Enamine

N-(3-methylphenyl)piperidin-4-amine

  • MDL number MFCD12822073
  • Cas Number : 1178145-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84072

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84072
Availability details for
Quantity
  • Availability
  • Add to Cart