ENA333559432 Enamine

methyl 2-chloro-4-fluoro-5-nitrobenzoate

  • MDL number MFCD07779372
  • Cas Number : 85953-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55-60
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84114

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84114
Availability details for
Quantity
  • Availability
  • Add to Cart