ENA333559437 Enamine

8-bromo-4-chloroquinoline-2-carboxylic acid

  • MDL number MFCD06810536
  • Cas Number : 902742-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84120

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84120
Availability details for
Quantity
  • Availability
  • Add to Cart