ENA333559440 Enamine

2-(3-acetylphenyl)-1lambda6,2-thiazolidine-1,1-dione

  • MDL number MFCD16320758
  • Cas Number : 1225595-21-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-84123

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84123
Availability details for
Quantity
  • Availability
  • Add to Cart