ENA333559444 Enamine

1-(3-methoxyphenyl)ethane-1-thiol

  • MDL number MFCD10691186
  • Cas Number : 1092300-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-84130

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84130
Availability details for
Quantity
  • Availability
  • Add to Cart