ENA333559452 Enamine

1-(2-fluorophenyl)-2-phenylethan-1-amine

  • MDL number MFCD09887496
  • Cas Number : 959407-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-84143

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84143
Availability details for
Quantity
  • Availability
  • Add to Cart