ENA333559453 Enamine

(heptan-4-yl)({8-methyl-8-azabicyclo[3.2.1]octan-3-yl}methyl)amine

  • MDL number MFCD12852300
  • Cas Number : 1178501-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-84144

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84144
Availability details for
Quantity
  • Availability
  • Add to Cart