ENA333559458 Enamine

(cyclobutylmethyl)(1-phenylethyl)amine

  • MDL number MFCD03211343
  • Cas Number : 356539-85-0
  • Molecular Weight : 189.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-84149

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84149
Availability details for
Quantity
  • Availability
  • Add to Cart