ENA333559464 Enamine

2-(cyclopropylmethoxy)aniline

  • MDL number MFCD08699323
  • Cas Number : 89523-17-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84176

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84176
Availability details for
Quantity
  • Availability
  • Add to Cart