ENA333559467 Enamine

2-(3,5-dimethylphenyl)ethan-1-amine

  • MDL number MFCD06212747
  • Cas Number : 76935-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84181

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84181
Availability details for
Quantity
  • Availability
  • Add to Cart