ENA333559473 Enamine

6-bromo-1-(2-methoxyethyl)-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD11148043
  • Cas Number : 1020994-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-84199

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84199
Availability details for
Quantity
  • Availability
  • Add to Cart