ENA333559475 Enamine

5-(4-bromophenyl)-4-methylthiophene-2-carboxylic acid

  • MDL number MFCD01830265
  • Cas Number : 62404-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83735

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83735
Availability details for
Quantity
  • Availability
  • Add to Cart